missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Alanine 4-nitroanilide hydrochloride, 98%, Thermo Scientific Chemicals
$76.70 - $675.68
Chemical Identifiers
CAS | 31796-55-1 |
---|---|
Molecular Formula | C9H12ClN3O3 |
Molecular Weight (g/mol) | 245.663 |
MDL Number | MFCD00039088 |
InChI Key | YEXRLSXNWLNHQR-RGMNGODLSA-N |
Synonym | h-ala-pna hcl, l-alanine 4-nitroanilide hydrochloride, h-ala-pna.hcl, alanine-p-nitroanilide hydrochloride, s-2-amino-n-4-nitrophenyl propionamide hydrochloride, s-2-amino-n-4-nitrophenyl propanamide hydrochloride, 2s-2-amino-n-4-nitrophenyl propanamide hydrochloride, h-ala-pna-hcl, h-l-ala-pna hcl, pubchem12864 |
PubChem CID | 2802426 |
IUPAC Name | (2S)-2-amino-N-(4-nitrophenyl)propanamide;hydrochloride |
SMILES | CC(C(=O)NC1=CC=C(C=C1)[N+](=O)[O-])N.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH63393MD
|
Thermo Scientific Chemicals
H63393MD |
250 mg |
Each for $76.70
|
|
||||||
AAH6339303
|
Thermo Scientific Chemicals
H6339303 |
1 g |
Each for $189.92
|
|
||||||
AAH6339306
|
Thermo Scientific Chemicals
H6339306 |
5 g |
Each for $675.68
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
31796-55-1 | |
245.663 | |
YEXRLSXNWLNHQR-RGMNGODLSA-N | |
2802426 | |
CC(C(=O)NC1=CC=C(C=C1)[N+](=O)[O-])N.Cl |
C9H12ClN3O3 | |
MFCD00039088 | |
h-ala-pna hcl, l-alanine 4-nitroanilide hydrochloride, h-ala-pna.hcl, alanine-p-nitroanilide hydrochloride, s-2-amino-n-4-nitrophenyl propionamide hydrochloride, s-2-amino-n-4-nitrophenyl propanamide hydrochloride, 2s-2-amino-n-4-nitrophenyl propanamide hydrochloride, h-ala-pna-hcl, h-l-ala-pna hcl, pubchem12864 | |
(2S)-2-amino-N-(4-nitrophenyl)propanamide;hydrochloride |
Specifications
31796-55-1 | |
C9H12ClN3O3 | |
6303731 | |
YEXRLSXNWLNHQR-RGMNGODLSA-N | |
(2S)-2-amino-N-(4-nitrophenyl)propanamide;hydrochloride | |
245.663 | |
245.66 | |
Powder |
Yellow | |
MFCD00039088 | |
h-ala-pna hcl, l-alanine 4-nitroanilide hydrochloride, h-ala-pna.hcl, alanine-p-nitroanilide hydrochloride, s-2-amino-n-4-nitrophenyl propionamide hydrochloride, s-2-amino-n-4-nitrophenyl propanamide hydrochloride, 2s-2-amino-n-4-nitrophenyl propanamide hydrochloride, h-ala-pna-hcl, h-l-ala-pna hcl, pubchem12864 | |
CC(C(=O)NC1=CC=C(C=C1)[N+](=O)[O-])N.Cl | |
250 mg | |
2802426 | |
98% | |
L-Alanine 4-nitroanilide hydrochloride |
Safety and Handling
EINECSNumber : 250-811-0
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only