missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Alanine tert-Butyl Ester Hydrochloride 97.0+%, TCI America™
Supplier: TCI America A20105G
Specifications
L-Alanine tert-Butyl Ester Hydrochloride | |
208°C | |
C7H16ClNO2 | |
5 g | |
WIQIWPPQGWGVHD-JEDNCBNOSA-N | |
tert-butyl (2S)-2-aminopropanoate hydrochloride | |
11275390 | |
≥97.0% (N) |
13404-22-3 | |
White | |
MFCD00035524 | |
h-ala-otbu.hcl, tert-butyl l-alaninate hydrochloride, l-alanine tert-butyl ester hydrochloride, s-tert-butyl 2-aminopropanoate hydrochloride, tert-butyl alaninate hydrochloride, h-ala-otbu inverted exclamation mark currencyhcl, tert-butyl 2s-2-aminopropanoate hydrochloride, l-alanine t-butyl ester hydrochloride, alanine, 1,1-dimethylethyl ester, hydrochloride, l-alanine, 1,1-dimethylethyl ester, hydrochloride | |
Cl.C[C@H](N)C(=O)OC(C)(C)C | |
181.66 | |
181.66 | |
Crystalline Powder |
Chemical Identifiers
13404-22-3 | |
181.66 | |
WIQIWPPQGWGVHD-JEDNCBNOSA-N | |
11275390 | |
Cl.C[C@H](N)C(=O)OC(C)(C)C |
C7H16ClNO2 | |
MFCD00035524 | |
h-ala-otbu.hcl, tert-butyl l-alaninate hydrochloride, l-alanine tert-butyl ester hydrochloride, s-tert-butyl 2-aminopropanoate hydrochloride, tert-butyl alaninate hydrochloride, h-ala-otbu inverted exclamation mark currencyhcl, tert-butyl 2s-2-aminopropanoate hydrochloride, l-alanine t-butyl ester hydrochloride, alanine, 1,1-dimethylethyl ester, hydrochloride, l-alanine, 1,1-dimethylethyl ester, hydrochloride | |
tert-butyl (2S)-2-aminopropanoate hydrochloride |
Safety and Handling
TSCA : No