missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(+)-Arginine Hydrochloride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 105000025

 View more versions of this product

Catalog No. AC105000025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(+)-Arginine hydrochloride
226.0°C
0.5% max. (105°C, 2 hrs)
98+%
C6H14N4O2·HCl
MFCD00064550
15,77
+ 22
Solubility in water: 75.1g/100g (20°C)
C(CC(C(=O)O)N)CN=C(N)N.Cl
95% min. (c=10, H2O, 430nm) 1cm cell
66250
98+%
1119-34-2
White
Authentic
Plastic bottle
H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
2.5 kg
+22° (20°C c=8,6N HCl)
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
KWTQSFXGGICVPE-WCCKRBBISA-N
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
210.67
210.67
Crystalline Powder or Crystals

Chemical Identifiers

1119-34-2
210.67
KWTQSFXGGICVPE-WCCKRBBISA-N
66250
C(CC(C(=O)O)N)CN=C(N)N.Cl
C6H14N4O2·HCl
MFCD00064550
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-275-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only