missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L(+)-Arginine Hydrochloride, 98+%, Thermo Scientific Chemicals
$36.43 - $917.96
Chemical Identifiers
CAS | 1119-34-2 |
---|---|
Molecular Formula | C6H14N4O2·HCl |
Molecular Weight (g/mol) | 210.67 |
MDL Number | MFCD00064550 |
InChI Key | KWTQSFXGGICVPE-WCCKRBBISA-N |
Synonym | l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y |
PubChem CID | 66250 |
IUPAC Name | (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride |
SMILES | C(CC(C(=O)O)N)CN=C(N)N.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC105000250
|
Thermo Scientific Chemicals
105000250 |
25 g | Plastic bottle |
N/A
|
|
||||
AC105005000
|
Thermo Scientific Chemicals
105005000 |
500 g | Plastic bottle |
N/A
|
|
||||
AC105000025
|
Thermo Scientific Chemicals
105000025 |
2.5 kg | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1119-34-2 | |
210.67 | |
KWTQSFXGGICVPE-WCCKRBBISA-N | |
66250 | |
C(CC(C(=O)O)N)CN=C(N)N.Cl |
C6H14N4O2·HCl | |
MFCD00064550 | |
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y | |
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride |
Specifications
1119-34-2 | |
White | |
Authentic | |
Plastic bottle | |
H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl | |
25 g | |
+ 22 | |
Solubility in water: 75.1g/100g (20°C) | |
C(CC(C(=O)O)N)CN=C(N)N.Cl | |
95% min. (c=10, H2O, 430nm) 1cm cell | |
66250 | |
98+% | |
L(+)-Arginine hydrochloride |
226°C | |
0.5% max. (105°C, 2 hrs) | |
98+% | |
C6H14N4O2·HCl | |
MFCD00064550 | |
15,77 | |
l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y | |
KWTQSFXGGICVPE-WCCKRBBISA-N | |
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride | |
210.67 | |
210.67 | |
Crystalline Powder or Crystals |
Safety and Handling
EINECSNumber : 214-275-1
RUO – Research Use Only