missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(+)-Ascorbic Acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 105021000

 View more versions of this product

Catalog No. AC105021000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(+)-Ascorbic acid
50-81-7
White
Authentic
Plastic bottle
MFCD00064328
18, IV, 3038
+ 20.50
(5% in water) Passes test
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
176.12
CHEBI:29073
99%
99%
190.0°C to 192.0°C
2.1 to 2.6 (5% aq. soln.)
98.5% min. (Iodimetry)
C6H8O6
100 g
15, 819
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CIWBSHSKHKDKBQ-DOMZIZNONA-N
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
54670067
176.13
Crystalline Powder or Crystals

Chemical Identifiers

50-81-7
176.12
CIWBSHSKHKDKBQ-DOMZIZNONA-N
54670067
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
C6H8O6
MFCD00064328
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CHEBI:29073
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
Videos
Safety and Handling

Safety and Handling

AIR SENSITIVE

LIGHT SENSITIVE

EINECSNumber : 200-066-2

RTECSNumber : CI7650000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only