missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(+)-Ascorbic Acid, 99%, Thermo Scientific Chemicals

$59.87 - $582.06

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC105021000
SDS View Documents
Thermo Scientific Chemicals
105021000
100 g Plastic bottle
Each for $59.87
 
AC105025000
SDS View Documents
Thermo Scientific Chemicals
105025000
500 g Plastic bottle
Each for $129.63
 
AC105020025
SDS View Documents
Thermo Scientific Chemicals
105020025
2.5 kg Plastic bottle
Each for $582.06
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-81-7
176.12
CIWBSHSKHKDKBQ-DOMZIZNONA-N
54670067
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
C6H8O6
MFCD00064328
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CHEBI:29073
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

50-81-7
White
Authentic
Plastic bottle
MFCD00064328
18, IV, 3038
+ 20.50
(5% in water) Passes test
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
176.12
CHEBI:29073
99%
L(+)-Ascorbic acid, 99%
190.0°C to 192.0°C
2.1 to 2.6 (5% aq. soln.)
98.5% min. (Iodimetry)
C6H8O6
100 g
15, 819
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
CIWBSHSKHKDKBQ-DOMZIZNONA-N
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
54670067
176.13
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

AIR SENSITIVE

LIGHT SENSITIVE

EINECSNumber : 200-066-2

RTECSNumber : CI7650000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only