missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Asparagine, 99% [Anhydrous], MP Biomedicals
Supplier: MP Biomedicals Inc 0219463080
Specifications
L-Asparagine, 99% [Anhydrous] | |
C4H8N2O3 | |
DCXYFEDJOCDNAF-REOHCLBHSA-N | |
(2S)-2,4-diamino-4-oxobutanoic acid | |
132.119 | |
CHEBI:17196 | |
Cell Culture |
70-47-3 | |
l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid | |
C(C(C(=O)O)N)C(=O)N | |
100 g | |
6267 | |
132.1 | |
Crystals |
Chemical Identifiers
70-47-3 | |
132.119 | |
l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid | |
CHEBI:17196 | |
C(C(C(=O)O)N)C(=O)N |
C4H8N2O3 | |
DCXYFEDJOCDNAF-REOHCLBHSA-N | |
6267 | |
(2S)-2,4-diamino-4-oxobutanoic acid |
Safety and Handling
Recommended Storage : Room temperature