missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Asparagine, 99% [Anhydrous], MP Biomedicals
$182.25 - $182.25
Chemical Identifiers
CAS | 70-47-3 |
---|---|
Molecular Formula | C4H8N2O3 |
Molecular Weight (g/mol) | 132.119 |
InChI Key | DCXYFEDJOCDNAF-REOHCLBHSA-N |
Synonym | l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid |
PubChem CID | 6267 |
ChEBI | CHEBI:17196 |
IUPAC Name | (2S)-2,4-diamino-4-oxobutanoic acid |
SMILES | C(C(C(=O)O)N)C(=O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
ICN19463080
|
MP Biomedicals Inc
0219463080 |
100 g |
Each for $182.25
|
|
||||||
Chemical Identifiers
70-47-3 | |
132.119 | |
l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid | |
CHEBI:17196 | |
C(C(C(=O)O)N)C(=O)N |
C4H8N2O3 | |
DCXYFEDJOCDNAF-REOHCLBHSA-N | |
6267 | |
(2S)-2,4-diamino-4-oxobutanoic acid |
Specifications
70-47-3 | |
l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid | |
C(C(C(=O)O)N)C(=O)N | |
100 g | |
6267 | |
132.1 | |
Crystals |
C4H8N2O3 | |
DCXYFEDJOCDNAF-REOHCLBHSA-N | |
(2S)-2,4-diamino-4-oxobutanoic acid | |
132.119 | |
CHEBI:17196 | |
Cell Culture | |
L-Asparagine, 99% [Anhydrous] |
Safety and Handling
Recommended Storage : Room temperature