missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Cysteine hydrochloride, 98%, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 434850250

 View more versions of this product

Catalog No. AC434850250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Cysteine hydrochloride
175.0°C to 181.0°C
22.29 to 23.43 % C 8.67 to 9.11 % N
98%
C3H8ClNO2S
MFCD00064553
15,2778
l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous
IFQSXNOEEPCSLW-QYLAHFPANA-N
(2R)-2-amino-3-sulfanylpropanoic acid;hydrochloride
60960
98%
52-89-1
White to Brown
Authentic
Glass bottle
HSCH2CH(NH2)CO2H·HCl
25 g
+ 7.00
Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone
Cl.N[C@@H](CS)C(O)=O
157.61
157.61
Crystalline Powder

Chemical Identifiers

52-89-1
157.61
IFQSXNOEEPCSLW-QYLAHFPANA-N
60960
Cl.N[C@@H](CS)C(O)=O
C3H8ClNO2S
MFCD00064553
l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous
(2R)-2-amino-3-sulfanylpropanoic acid;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-157-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only