missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Cysteine hydrochloride, 98%, anhydrous, Thermo Scientific Chemicals

$168.28 - $3483.13

Chemical Identifiers

CAS 52-89-1
Molecular Formula C3H8ClNO2S
Molecular Weight (g/mol) 157.61
MDL Number MFCD00064553
InChI Key IFQSXNOEEPCSLW-QYLAHFPANA-N
Synonym l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous
PubChem CID 60960
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrochloride
SMILES Cl.N[C@@H](CS)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434850250
SDS View Documents
Thermo Scientific Chemicals
434850250
25 g Glass bottle
Each for $168.28
 
AC434851000
SDS View Documents
Thermo Scientific Chemicals
434851000
100 g Glass bottle
Each for $387.90
 
AC434850010
SDS View Documents
Thermo Scientific Chemicals
434850010
1 kg Glass bottle
Each for $3,483.13
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

52-89-1
157.61
IFQSXNOEEPCSLW-QYLAHFPANA-N
60960
Cl.N[C@@H](CS)C(O)=O
C3H8ClNO2S
MFCD00064553
l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous
(2R)-2-amino-3-sulfanylpropanoic acid;hydrochloride

Specifications

52-89-1
White to Brown
Authentic
Glass bottle
HSCH2CH(NH2)CO2H·HCl
25 g
+ 7.00
Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone
Cl.N[C@@H](CS)C(O)=O
157.61
157.61
Crystalline Powder
175.0°C to 181.0°C
22.29 to 23.43 % C 8.67 to 9.11 % N
98%
C3H8ClNO2S
MFCD00064553
15,2778
l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous
IFQSXNOEEPCSLW-QYLAHFPANA-N
(2R)-2-amino-3-sulfanylpropanoic acid;hydrochloride
60960
98%
L-Cysteine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-157-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only