missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate 98.0+%, TCI America™
Supplier: TCI America H1657100MG
Specifications
L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate | |
133°C | |
C19H26O13 | |
LD-Heptose Peracetate | |
≥98.0% (GC) |
104486-80-8 | |
White | |
100 mg | |
462.40 | |
Crystalline Powder |
Chemical Identifiers
104486-80-8 | |
C19H26O13 | |
LD-Heptose Peracetate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator