missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate 98.0+%, TCI America™
$100.35 - $297.57
Chemical Identifiers
CAS | 104486-80-8 |
---|---|
Molecular Formula | C19H26O13 |
Synonym | LD-Heptose Peracetate |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
H165720MG
|
TCI America
H165720MG |
20 mg |
Each for $100.35
|
|
|||||
H1657100MG
|
TCI America
H1657100MG |
100 mg |
Each for $297.57
|
|
|||||
Chemical Identifiers
104486-80-8 | |
C19H26O13 | |
LD-Heptose Peracetate |
Specifications
104486-80-8 | |
White | |
20 mg | |
462.40 | |
Crystalline Powder |
133°C | |
C19H26O13 | |
LD-Heptose Peracetate | |
≥98.0% (GC) | |
L-glycero-alpha-D-manno-Heptopyranose 1,2,3,4,6,7-Hexaacetate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator