missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals L-Histidine, 98%

Supplier:  Thermo Scientific Chemicals 166151000

 View more versions of this product

Catalog No. AC166151000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

282.0°C
200ppm max.
7.0 to 8.5 (2% aq. soln.)
200ppm max.
0.5% max. (TLC)
Authentic
0.2% max. (105°C, 3 hrs)
C6H9N3O2
MFCD00064315
25, 513
98% min. (c=5, H2O, 430nm) 1cm cell
0.1% max.
Solubility in water: 41.6g/L (25 c). Other solubilities: soluble in mineral acids, insoluble in organic solvents
NC(CC1=CN=CN1)C(O)=O
+12.40° (20°C c=11,6N HCl)
100 g
CHEBI:15971
98%
L-Histidine
71-00-1
White or Yellow
200ppm max.
10ppm max.
10ppm max.
98%
Plastic Bottle
+ 12.40
200ppm max.
15, 4758
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
HNDVDQJCIGZPNO-UHFFFAOYNA-N
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
155.16
6274
155.16
Powder, Lumpy Powder or Melt

Chemical Identifiers

71-00-1
155.16
HNDVDQJCIGZPNO-UHFFFAOYNA-N
6274
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
C6H9N3O2
MFCD00064315
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
CHEBI:15971
NC(CC1=CN=CN1)C(O)=O
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only