missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals L-Histidine, 98%
$72.62 - $711.99
Chemical Identifiers
CAS | 71-00-1 |
---|---|
Molecular Formula | C6H9N3O2 |
Molecular Weight (g/mol) | 155.16 |
MDL Number | MFCD00064315 |
InChI Key | HNDVDQJCIGZPNO-UHFFFAOYNA-N |
Synonym | l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin |
PubChem CID | 6274 |
ChEBI | CHEBI:15971 |
IUPAC Name | (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid |
SMILES | NC(CC1=CN=CN1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC166150250
|
Thermo Scientific Chemicals
166150250 |
25 g | Plastic Bottle |
Each for $72.62
|
|
|||||
AC166151000
|
Thermo Scientific Chemicals
166151000 |
100 g | Plastic Bottle |
Each for $196.07
|
|
|||||
AC166155000
|
Thermo Scientific Chemicals
166155000 |
500 g | Plastic Bottle |
Each for $711.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
71-00-1 | |
155.16 | |
HNDVDQJCIGZPNO-UHFFFAOYNA-N | |
6274 | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid |
C6H9N3O2 | |
MFCD00064315 | |
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin | |
CHEBI:15971 | |
NC(CC1=CN=CN1)C(O)=O |
Specifications
282.0°C | |
71-00-1 | |
White or Yellow | |
200ppm max. | |
10ppm max. | |
10ppm max. | |
98% | |
Plastic Bottle | |
+ 12.40 | |
200ppm max. | |
15, 4758 | |
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin | |
HNDVDQJCIGZPNO-UHFFFAOYNA-N | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid | |
155.16 | |
6274 | |
155.16 | |
Powder, Lumpy Powder or Melt |
200ppm max. | |
7.0 to 8.5 (2% aq. soln.) | |
200ppm max. | |
0.5% max. (TLC) | |
Authentic | |
0.2% max. (105°C, 3 hrs) | |
C6H9N3O2 | |
MFCD00064315 | |
25, 513 | |
98% min. (c=5, H2O, 430nm) 1cm cell | |
0.1% max. | |
Solubility in water: 41.6g/L (25 c). Other solubilities: soluble in mineral acids, insoluble in organic solvents | |
NC(CC1=CN=CN1)C(O)=O | |
+12.40° (20°C c=11,6N HCl) | |
25 g | |
CHEBI:15971 | |
98% | |
L-Histidine |
RUO – Research Use Only