missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Histidine Monohydrochloride Monohydrate, MP Biomedical
Supplier: MP Biomedicals Inc 0210195790
Specifications
L-Histidine Monohydrochloride Monohydrate | |
White | |
MFCD00151027 | |
CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride | |
209.63 | |
209.6 |
5934-29-2 | |
C6H12ClN3O3 | |
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
C1=C(NC=N1)CC(C(=O)O)N.O.Cl | |
500 g | |
165377 | |
Powder |
Chemical Identifiers
5934-29-2 | |
209.63 | |
CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
165377 | |
C1=C(NC=N1)CC(C(=O)O)N.O.Cl |
C6H12ClN3O3 | |
MFCD00151027 | |
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C).