missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Lysine ethyl ester diisocyanate, 97%, Thermo Scientific Chemicals

$239.23 - $962.40

Chemical Identifiers

CAS 45172-15-4
Molecular Formula C10H14N2O4
Molecular Weight (g/mol) 226.23
MDL Number MFCD08276480
InChI Key IGWTZHMYJZZIGC-IMWMWJONNA-N
Synonym s-ethyl 2,6-diisocyanatohexanoate, l-lysine ethyl ester diisocyanate, ethyllysine diisocyanate, ethyllysine diisocyanate;, ksc005q3n, lysine ethyl ester diisocyanate;, l-lysine diisocyanate ethyl ester, l-lysine ethyl ester diisocyanate;, ethyl 2s-2,6-diisocyanatohexanoate, s-2,6-diisocyanatohexanoic acid ethyl ester
PubChem CID 71440643
IUPAC Name ethyl (2S)-2,6-diisocyanatohexanoate
SMILES CCOC(=O)[C@H](CCCCN=C=O)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6438406
View Documents
Thermo Scientific Chemicals
H6438406
5 g
Each for $239.23
 
AAH6438414
View Documents
Thermo Scientific Chemicals
H6438414
25 g
Each for $962.40
 
Description

Description

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Sparingly soluble in water (0.19 g/L).

Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Specifications

Chemical Identifiers

45172-15-4
226.23
IGWTZHMYJZZIGC-IMWMWJONNA-N
71440643
CCOC(=O)[C@H](CCCCN=C=O)N=C=O
C10H14N2O4
MFCD08276480
s-ethyl 2,6-diisocyanatohexanoate, l-lysine ethyl ester diisocyanate, ethyllysine diisocyanate, ethyllysine diisocyanate;, ksc005q3n, lysine ethyl ester diisocyanate;, l-lysine diisocyanate ethyl ester, l-lysine ethyl ester diisocyanate;, ethyl 2s-2,6-diisocyanatohexanoate, s-2,6-diisocyanatohexanoic acid ethyl ester
ethyl (2S)-2,6-diisocyanatohexanoate

Specifications

45172-15-4
C10H14N2O4
MFCD08276480
Sparingly soluble in water (0.19g/L).
CCOC(=O)[C@H](CCCCN=C=O)N=C=O
226.23
71440643
97%
Viscous Liquid
Yellow to Brown
1.4572
s-ethyl 2,6-diisocyanatohexanoate, l-lysine ethyl ester diisocyanate, ethyllysine diisocyanate, ethyllysine diisocyanate;, ksc005q3n, lysine ethyl ester diisocyanate;, l-lysine diisocyanate ethyl ester, l-lysine ethyl ester diisocyanate;, ethyl 2s-2,6-diisocyanatohexanoate, s-2,6-diisocyanatohexanoic acid ethyl ester
IGWTZHMYJZZIGC-IMWMWJONNA-N
ethyl (2S)-2,6-diisocyanatohexanoate
5 g
226.23
Moisture sensitive
L-Lysine ethyl ester diisocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P311-P333+P313-P362-P501c

H315-H317-H319-H331-H334-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only