Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
missing translation for 'pleaseLogIn'
L(+)-Ornithine hydrochloride, 99%, Thermo Scientific Chemicals
$73.84 - $675.77
Chemical Identifiers
CAS | 3184-13-2 |
---|---|
Molecular Formula | C5H12N2O2·HCl |
Molecular Weight (g/mol) | 168.62 |
MDL Number | MFCD00064562 |
InChI Key | GGTYBZJRPHEQDG-WCCKRBBISA-N |
Synonym | l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate |
PubChem CID | 76654 |
IUPAC Name | (2S)-2,5-diaminopentanoic acid;hydrochloride |
SMILES | C(CC(C(=O)O)N)CN.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
AC129560250
|
Thermo Scientific Chemicals
129560250 |
25 g | Plastic bottle |
Each for $73.84
|
|
||||
AC129561000
|
Thermo Scientific Chemicals
129561000 |
100 g | Plastic bottle |
Each for $177.76
|
|
||||
AC129565000
|
Thermo Scientific Chemicals
129565000 |
500 g | Plastic bottle |
Each for $675.77
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3184-13-2 | |
168.62 | |
GGTYBZJRPHEQDG-WCCKRBBISA-N | |
76654 | |
C(CC(C(=O)O)N)CN.Cl |
C5H12N2O2·HCl | |
MFCD00064562 | |
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate | |
(2S)-2,5-diaminopentanoic acid;hydrochloride |
Specifications
3184-13-2 | |
White | |
98.5% min. (Argentometry) | |
C5H12N2O2·HCl | |
MFCD00064562 | |
04, 420 | |
+ 24.00 (20.00°C c=4,6N HCl) | |
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate | |
GGTYBZJRPHEQDG-WCCKRBBISA-N | |
(2S)-2,5-diaminopentanoic acid;hydrochloride | |
76654 | |
99% | |
L(+)-Ornithine hydrochloride |
245.0°C | |
Authentic | |
Plastic bottle | |
H2N(CH2)3CH(NH2)CO2H·HCl | |
25 g | |
14, 6874 | |
+ 24.00 | |
(5% in 1 N HCl) Clear colorless | |
C(CC(C(=O)O)N)CN.Cl | |
168.62 | |
168.62 | |
Powder |
Safety and Handling
EINECSNumber : 221-678-6
RUO – Research Use Only