missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(+)-Penicillamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 416870010

 View more versions of this product

Catalog No. AC416870010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(+)-Penicillamine
1113-41-3
White
98%
C5H11NO2S
MFCD00064303
04, III, 1662
+63° (20°C c=5,1 M NaOH)
l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine
VVNCNSJFMMFHPL-GSVOUGTGSA-N
(2R)-2-amino-3-methyl-3-sulfanylbutanoic acid
92863
149.21
Crystalline Powder
98%
206.0°C
Authentic
Glass bottle
(CH3)2C(SH)CH(NH2)CO2H
1 g
15, 7197
+ 63.00
(5% in water) Clear colorless
CC(C)(C(C(=O)O)N)S
149.21
CHEBI:50869
98%

Chemical Identifiers

1113-41-3
149.21
VVNCNSJFMMFHPL-GSVOUGTGSA-N
92863
(2R)-2-amino-3-methyl-3-sulfanylbutanoic acid
C5H11NO2S
MFCD00064303
l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine
CHEBI:50869
CC(C)(C(C(=O)O)N)S
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 214-203-9

RTECSNumber : YV9445500

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only