Learn More
L(+)-Penicillamine, 98%, Thermo Scientific Chemicals
$87.36 - $87.36
Chemical Identifiers
CAS | 1113-41-3 |
---|---|
Molecular Formula | C5H11NO2S |
Molecular Weight (g/mol) | 149.21 |
MDL Number | MFCD00064303 |
InChI Key | VVNCNSJFMMFHPL-GSVOUGTGSA-N |
Synonym | l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine |
PubChem CID | 92863 |
ChEBI | CHEBI:50869 |
IUPAC Name | (2R)-2-amino-3-methyl-3-sulfanylbutanoic acid |
SMILES | CC(C)(C(C(=O)O)N)S |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC416870010
|
Thermo Scientific Chemicals
416870010 |
1 g | Glass bottle |
Each for $87.36
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1113-41-3 | |
149.21 | |
VVNCNSJFMMFHPL-GSVOUGTGSA-N | |
92863 | |
(2R)-2-amino-3-methyl-3-sulfanylbutanoic acid |
C5H11NO2S | |
MFCD00064303 | |
l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine | |
CHEBI:50869 | |
CC(C)(C(C(=O)O)N)S |
Specifications
1113-41-3 | |
White | |
98% | |
C5H11NO2S | |
MFCD00064303 | |
04, III, 1662 | |
+63° (20°C c=5,1 M NaOH) | |
l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine | |
VVNCNSJFMMFHPL-GSVOUGTGSA-N | |
(2R)-2-amino-3-methyl-3-sulfanylbutanoic acid | |
92863 | |
149.21 | |
Crystalline Powder |
206.0°C | |
Authentic | |
Glass bottle | |
(CH3)2C(SH)CH(NH2)CO2H | |
1 g | |
15, 7197 | |
+ 63.00 | |
(5% in water) Clear colorless | |
CC(C)(C(C(=O)O)N)S | |
149.21 | |
CHEBI:50869 | |
98% | |
L(+)-Penicillamine, 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 214-203-9
RTECSNumber : YV9445500
RUO – Research Use Only