missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(-)-Perillaldehyde, 90%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 203660250

 View more versions of this product

Catalog No. AC203660250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(-)-Perillaldehyde
18031-40-8
0.9700g/mL
95°C
88% min
C10H14O
MFCD00001543
07, 158
14, 7167
-115
RUMOYJJNUMEFDD-SNVBAGLBSA-N
(4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde
2724159
150.22
Liquid
90%
Yellow
104°C to 105°C (10.0 mmHg)
Authentic
Glass bottle
1.5040 to 1.51
25 g
0.97
− 115.00 (20.00°C c=10,C2H5OH)
--perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s
CC(=C)C1CCC(=CC1)C=O
150.22
CHEBI:86938
90%

Chemical Identifiers

18031-40-8
150.22
RUMOYJJNUMEFDD-SNVBAGLBSA-N
2724159
(4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde
C10H14O
MFCD00001543
--perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s
CHEBI:86938
CC(=C)C1CCC(=CC1)C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

RTECSNumber : GW2967200

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only