Learn More
L(-)-Perillaldehyde, 90%, Thermo Scientific Chemicals
$60.89 - $159.04
Chemical Identifiers
CAS | 18031-40-8 |
---|---|
Molecular Formula | C10H14O |
Molecular Weight (g/mol) | 150.22 |
MDL Number | MFCD00001543 |
InChI Key | RUMOYJJNUMEFDD-SNVBAGLBSA-N |
Synonym | --perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s |
PubChem CID | 2724159 |
ChEBI | CHEBI:86938 |
IUPAC Name | (4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde |
SMILES | CC(=C)C1CCC(=CC1)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC203660050
|
Thermo Scientific Chemicals
203660050 |
5 g | Glass bottle |
Each for $60.89
|
|
||||
AC203660250
|
Thermo Scientific Chemicals
203660250 |
25 g | Glass bottle |
Each for $159.04
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18031-40-8 | |
150.22 | |
RUMOYJJNUMEFDD-SNVBAGLBSA-N | |
2724159 | |
(4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde |
C10H14O | |
MFCD00001543 | |
--perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s | |
CHEBI:86938 | |
CC(=C)C1CCC(=CC1)C=O |
Specifications
18031-40-8 | |
0.9700g/mL | |
95°C | |
88% min | |
C10H14O | |
MFCD00001543 | |
07, 158 | |
14, 7167 | |
-115 | |
RUMOYJJNUMEFDD-SNVBAGLBSA-N | |
(4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde | |
2724159 | |
150.22 | |
Liquid |
Yellow | |
104°C to 105°C (10.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5040 to 1.51 | |
5 g | |
0.97 | |
− 115.00 (20.00°C c=10,C2H5OH) | |
--perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s | |
CC(=C)C1CCC(=CC1)C=O | |
150.22 | |
CHEBI:86938 | |
90% | |
L(-)-Perillaldehyde, 90% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
RTECSNumber : GW2967200
RUO – Research Use Only