missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals

$83.23 - $191.74

Chemical Identifiers

CAS 868-14-4
Molecular Formula C4H5KO6
Molecular Weight (g/mol) 188.18
MDL Number MFCD00065392,MFCD00071626,MFCD00064206
InChI Key KYKNRZGSIGMXFH-ZVGUSBNCSA-M
Synonym potassium bitartrate, l +-potassium hydrogen tartrate, potassium 3-carboxy-2,3-dihydroxypropanoate, monopotassium tartrate, potassium acid tartrate, kaliumtartrat, potassium hydrogentartrate, 1-potassium hydrogentartrate, ksc657e1t, butanedioic acid, 2,3-dihydroxy-2r,3r-, potassium salt 1:1
PubChem CID 24193652
IUPAC Name 2,3-dihydroxybutanedioic acid;potassium
SMILES [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC222672500
SDS View Documents
Thermo Scientific Chemicals
222672500
250 g Plastic bottle
Each for $83.23
 
AC222670010
SDS View Documents
Thermo Scientific Chemicals
222670010
1 kg Plastic bottle
Each for $191.74
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

868-14-4
188.18
KYKNRZGSIGMXFH-ZVGUSBNCSA-M
24193652
[K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
C4H5KO6
MFCD00065392,MFCD00071626,MFCD00064206
potassium bitartrate, l +-potassium hydrogen tartrate, potassium 3-carboxy-2,3-dihydroxypropanoate, monopotassium tartrate, potassium acid tartrate, kaliumtartrat, potassium hydrogentartrate, 1-potassium hydrogentartrate, ksc657e1t, butanedioic acid, 2,3-dihydroxy-2r,3r-, potassium salt 1:1
2,3-dihydroxybutanedioic acid;potassium

Specifications

868-14-4
1.9500g/mL
Authentic
Plastic bottle
KO2CCH(OH)CH(OH)CO2H
250 g
1.95
+ 32.50 (20.00°C c=10,1N NaOH)
potassium bitartrate, l +-potassium hydrogen tartrate, potassium 3-carboxy-2,3-dihydroxypropanoate, monopotassium tartrate, potassium acid tartrate, kaliumtartrat, potassium hydrogentartrate, 1-potassium hydrogentartrate, ksc657e1t, butanedioic acid, 2,3-dihydroxy-2r,3r-, potassium salt 1:1
[K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
188.18
188.18
Crystals or Powder
Colorless or White
0.5% max. (105°C)
99%
C4H5KO6
MFCD00065392,MFCD00071626,MFCD00064206
03, 529
15, 7736
+ 32.50
KYKNRZGSIGMXFH-ZVGUSBNCSA-M
2,3-dihydroxybutanedioic acid;potassium
24193652
99%
L(+)-Potassium hydrogen tartrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-769-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only