missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Prolinamide, 98%, Thermo Scientific™

Chemical Identifiers

CAS 7531-52-4
Molecular Formula C5H11N2O
Molecular Weight (g/mol) 115.16
MDL Number MFCD00005253
InChI Key VLJNHYLEOZPXFW-BYPYZUCNSA-O
Synonym l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
PubChem CID 111306
ChEBI CHEBI:21374
IUPAC Name (2S)-pyrrolidine-2-carboxamide
SMILES NC(=O)[C@@H]1CCC[NH2+]1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC302990010
SDS View Documents
Thermo Scientific Chemicals
302990010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC302990050
SDS View Documents
Thermo Scientific Chemicals
302990050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7531-52-4
115.16
VLJNHYLEOZPXFW-BYPYZUCNSA-O
111306
(2S)-pyrrolidine-2-carboxamide
C5H11N2O
MFCD00005253
l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
CHEBI:21374
NC(=O)[C@@H]1CCC[NH2+]1

Specifications

7531-52-4
Authentic
C5H11N2O
MFCD00005253
l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
NC(=O)[C@@H]1CCC[NH2+]1
−106° (20°C c=2,EtOH)
115.16
CHEBI:21374
98%
97.5% min. (TLC)
98%
Glass bottle
− 106.00
VLJNHYLEOZPXFW-BYPYZUCNSA-O
(2S)-pyrrolidine-2-carboxamide
1g
111306
114.15
L-Prolinamide
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only