missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Proline, Cell Culture Reagent, ≥99%, MP Biomedicals™

L-Proline, Cell Culture Reagent, ≥99% is used in cell culture media and as a component of MEM non-essential amino acids solution.

Supplier:  MP Biomedicals Inc 0219472880

 View more versions of this product

Catalog No. ICN19472880


Explore available promotions

Description

Description

  • L-Proline is used in cell culture media.
  • L-Proline is also a component of MEM non-essential amino acids solution.
Specifications

Specifications

221°C
Hydroxy-L-proline free
C5H9NO2
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
ONIBWKKTOPOVIA-UHFFFAOYNA-N
1.064 g/mL at 24°C
100 g
145742
-82° to -85° (c=1.6, water)
≥99%
Powder
L-Proline
147-85-3
MFCD00064318
Soluble in water, 162,000 mg/L at 25°C
OC(=O)C1CCCN1
pyrrolidine-2-carboxylic acid
115.13
CHEBI:17203
115.1
Cell Culture Reagent
1.95 (COOH), 10.64 (NH2)

Chemical Identifiers

147-85-3
115.13
ONIBWKKTOPOVIA-UHFFFAOYNA-N
145742
pyrrolidine-2-carboxylic acid
C5H9NO2
MFCD00064318
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh
CHEBI:17203
OC(=O)C1CCCN1
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room temperature

SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.