missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Proline, Cell Culture Reagent, ≥99%, MP Biomedicals™
L-Proline, Cell Culture Reagent, ≥99% is used in cell culture media and as a component of MEM non-essential amino acids solution.
$290.75 - $290.75
Chemical Identifiers
CAS | 147-85-3 |
---|---|
Molecular Formula | C5H9NO2 |
Molecular Weight (g/mol) | 115.13 |
MDL Number | MFCD00064318 |
InChI Key | ONIBWKKTOPOVIA-UHFFFAOYNA-N |
Synonym | l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh |
PubChem CID | 145742 |
ChEBI | CHEBI:17203 |
IUPAC Name | pyrrolidine-2-carboxylic acid |
SMILES | OC(=O)C1CCCN1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN19472880
|
MP Biomedicals Inc
0219472880 |
100 g |
Each for $290.75
|
|
|||||
Description
- L-Proline is used in cell culture media.
- L-Proline is also a component of MEM non-essential amino acids solution.
Chemical Identifiers
147-85-3 | |
115.13 | |
ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
145742 | |
pyrrolidine-2-carboxylic acid |
C5H9NO2 | |
MFCD00064318 | |
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
CHEBI:17203 | |
OC(=O)C1CCCN1 |
Specifications
221°C | |
C5H9NO2 | |
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
1.064 g/mL at 24°C | |
100 g | |
145742 | |
-82° to -85° (c=1.6, water) | |
≥99% | |
Powder | |
L-Proline, Hydroxy-L-proline free |
147-85-3 | |
MFCD00064318 | |
Soluble in water, 162,000 mg/L at 25°C | |
OC(=O)C1CCCN1 | |
pyrrolidine-2-carboxylic acid | |
115.13 | |
CHEBI:17203 | |
115.1 | |
Cell Culture Reagent | |
1.95 (COOH), 10.64 (NH2) |
Safety and Handling
Recommended Storage : Room temperature
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.