missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Proline, >99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210273091
Specifications
L-Proline | |
147-85-3 | |
>99% | |
MFCD00064318 | |
ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
pyrrolidine-2-carboxylic acid | |
115.13 | |
CHEBI:17203 | |
>99% |
Cell Biology and Analysis | |
White | |
C5H9NO2 | |
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
OC(=O)C1CCCN1 | |
1 kg | |
145742 | |
115.13 | |
Powder |
Chemical Identifiers
147-85-3 | |
115.13 | |
ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
145742 | |
pyrrolidine-2-carboxylic acid |
C5H9NO2 | |
MFCD00064318 | |
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
CHEBI:17203 | |
OC(=O)C1CCCN1 |
Safety and Handling
EINECSNumber : 205-702-2
Recommended Storage : Store at Room Temperature.