missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-threonine methyleester hydrochloride, +98%, Thermo Scientific™
Chemical Identifiers
CAS | 39994-75-7 |
---|---|
Molecular Formula | C5H12NO3 |
Molecular Weight (g/mol) | 134.15 |
MDL Number | MFCD00037677 |
InChI Key | TVHCXXXXQNWQLP-DMTCNVIQSA-O |
Synonym | l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl |
PubChem CID | 2734893 |
IUPAC Name | methyl (2S,3R)-2-amino-3-hydroxybutanoate;hydrochloride |
SMILES | COC(=O)[C@@H]([NH3+])[C@@H](C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC376690010
|
Thermo Scientific Chemicals
376690010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC376690050
|
Thermo Scientific Chemicals
376690050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC376690250
|
Thermo Scientific Chemicals
376690250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
39994-75-7 | |
134.15 | |
TVHCXXXXQNWQLP-DMTCNVIQSA-O | |
2734893 | |
COC(=O)[C@@H]([NH3+])[C@@H](C)O |
C5H12NO3 | |
MFCD00037677 | |
l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl | |
methyl (2S,3R)-2-amino-3-hydroxybutanoate;hydrochloride |
Specifications
39994-75-7 | |
C5H12NO3 | |
CH3CH(OH)CH(NH2)C(O)OCH3·HCl | |
Glass bottle | |
TVHCXXXXQNWQLP-DMTCNVIQSA-O | |
methyl (2S,3R)-2-amino-3-hydroxybutanoate;hydrochloride | |
134.15 | |
169.61 | |
1% Max. | |
1g |
98+% | |
Authentic | |
MFCD00037677 | |
l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl | |
COC(=O)[C@@H]([NH3+])[C@@H](C)O | |
-9.5° to -10.5° (20°C, 589nm) (c=1, Methanol, 20°C) | |
2734893 | |
≥98.0% ((on dry substance)) | |
−10° (20°C c=1,methanol) | |
L-Threonine methyleester hydrochloride |
RUO â Research Use Only