missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Tryptophan, ≥99%, MP Biomedicals™

L-Tryptophan is an amino acid precursor of serotonin and melatonin

Supplier:  MP Biomedicals Inc 0210315125

 View more versions of this product

Catalog No. ICN10315125


Explore available promotions

Description

Description

Specifications

Specifications

L-Tryptophan
White
C11H12N2O2
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
25 g
6305
204.22
73-22-3
≥99%
MFCD00064340
QIVBCDIJIAJPQS-VIFPVBQESA-N
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
204.23
CHEBI:16828
Crystalline Powder

Chemical Identifiers

73-22-3
204.23
QIVBCDIJIAJPQS-VIFPVBQESA-N
6305
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
C11H12N2O2
MFCD00064340
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
CHEBI:16828
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-795-6

Recommended Storage : Store at Room temperature (15 to 30°C).

SDS
Documents

Documents

Promotions

Promotions