missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Tryptophan, ≥99%, MP Biomedicals™

L-Tryptophan is an amino acid precursor of serotonin and melatonin

$65.19 - $1141.36

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10315125
View Documents
MP Biomedicals Inc
0210315125
25 g
Each for $65.19
 
ICN10315180
View Documents
MP Biomedicals Inc
0210315180
100 g
Each for $193.76
 
ICN10315190
View Documents
MP Biomedicals Inc
0210315190
500 g
Each for $762.00
 
ICN10315191
View Documents
MP Biomedicals Inc
0210315191
1 kg
Each for $1,141.36
 
Description

Description

Specifications

Chemical Identifiers

73-22-3
204.23
QIVBCDIJIAJPQS-VIFPVBQESA-N
6305
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
C11H12N2O2
MFCD00064340
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
CHEBI:16828
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

73-22-3
≥99%
MFCD00064340
QIVBCDIJIAJPQS-VIFPVBQESA-N
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
204.23
CHEBI:16828
Crystalline Powder
White
C11H12N2O2
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
25 g
6305
204.22
L-Tryptophan
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-795-6

Recommended Storage : Store at Room temperature (15 to 30°C).

SDS
missing translation for 'documents'

missing translation for 'documents'