missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Tryptophan, ≥99%, MP Biomedicals™
L-Tryptophan is an amino acid precursor of serotonin and melatonin
Supplier: MP Biomedicals Inc 0210315190
Specifications
L-Tryptophan | |
White | |
C11H12N2O2 | |
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin | |
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O | |
500 g | |
6305 | |
204.22 |
73-22-3 | |
≥99% | |
MFCD00064340 | |
QIVBCDIJIAJPQS-VIFPVBQESA-N | |
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid | |
204.23 | |
CHEBI:16828 | |
Crystalline Powder |
Chemical Identifiers
73-22-3 | |
204.23 | |
QIVBCDIJIAJPQS-VIFPVBQESA-N | |
6305 | |
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid |
C11H12N2O2 | |
MFCD00064340 | |
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin | |
CHEBI:16828 | |
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O |
Safety and Handling
EINECSNumber : 200-795-6
Recommended Storage : Store at Room temperature (15 to 30°C).