Fisherbrand™ Lab Grade Cylindrical Dewar Flasks with Plastic Mesh Casing
Borosilicate glass with silvered double walls and mesh casing for better grip
Supplier:  Fisherbrand™ FB10197C
 View more versions of this product
Catalog No. FB10197C
Explore available promotions
Description
Description
  • Cylindrical- Wide Mouth
  • Borosilicate Glass
  • Silvered Double Walls
  • PE Stopper Included
  • For Use with Liquified Gases
  • Aluminum Housing
  • Lab Grade
  • Plastic Mesh Casing
Specifications
Specifications
Plastic Mesh Casing
4.75 in.
256 mm
Silver
1900 mL
120 mm
Liquid Nitrogen
190 mm
Aluminum