Labconco™ 6 Ft. Protector™ Premier™ Laboratory Hood
Choose a Labconco Protector™ Premier™ Laboratory Hood to outfit your lab when working with general chemistry applications.
Supplier:  Labconco™ 100600000
 View more versions of this product
Catalog No. 10-369-134
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Incorporate a sleek interior with a molded one-piece fiberglass liner
  • Specially formulated, fiberglass-reinforced polyester offers corrosion, fire resistance and easy clean up
  • Hoods are suitable for general chemistry applications
  • Incorporate many containment-enhancing features including Clean-Sweep™ technology and Eco-Foil™ air foil
  • Testing confirms the Protector Premier Hood meets the SEFA-1 standard of a low velocity, high performance hood and may be operated as low as 60 fpm
  • Offered for use with a remotely located blower or with a built-in blower
Specifications
Specifications
Laboratory Fume Hood
LED
10 A
31.7 in.
Hood w/o Services and Blower
59 in.
149.9 cm
100/115 V
220 kg
182.9 cm
Powder
Powder-coated Steel
ANSI Z9.5, ASHRAE 110, ASTM E84, CAN/CSA C22.2, NFPA 45, SEFA 1, UL
80.5 cm
735 cfm
18 in.
Fiberglass
485 lb.
72 in.