Labconco™ Fume Hood Standard Service Fixture Kits (Fiberglass 30 or Basic Hoods)
Keep your Labconco fume hood in top operation with a Fume Hood Standard Service Fixture Kit, available in several types to meet your specific needs.
Supplier: Labconco™ 2832601
Description
- Remote controlled
- Right-side mount
- Fits Fiberglass 30 or Basic hoods
- Available types: Air, Argon, Cold Water, Deionized/Distilled Water, Gas, Hot Water, Nitrogen, Steam, Vacuum
Specifications
Hot Water Service Fixture Kit | |
Fiberglass 30 Laboratory Hood | |
40 psi | |
1.8 kg |
3.5 gpm | |
Remotely controlled service fixture with 0.25 in. dia. brass valve and copper tubing, red fixture knob and red hose connector, mounting on right side of fume hoods | |
4 lb. |