Labconco™ Guardian™ Digital Airflow Monitor: For Premier, Protector
Monitor and alert operator to low airflow conditions
Supplier:  Labconco™ 9413401
 View more versions of this product
Catalog No. 10-369-207
Explore available promotions
Includes: Faceplate with circuit board, electrical power pack, vinyl tubing, side wall flow tube adapter, and wiring instructions
Description
Description
Monitor and alert operator to low airflow conditions
If hood face velocity falls below programmable setpoint, audible alarm sounds and LOW indicator glows red. From faceplate, user can easily select and calibrate a setpoint between 50 and 250fpm using a velocity meter.
  • LCD displays actual face velocity
  • TEST RESET switch mutes audible alarm
  • EMERG. PURGE switch can be used to activate a relay to open hood damper (not included)
Order Info
Installation requires factory-prepared cut-out in upper portion of front panel (for ClassMate) or right fixture panel (for other models)
Specifications
Specifications
Premier, Protector XL, Protector Xstream Lab Hoods
208/230 V
208 to 230 V, 50/60 Hz
50/60 Hz