missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Purifier™ Vertical Clean Bench

Protect work from particulate contamination by choosing a Purifier™ Vertical Clean Bench.

Supplier:  Labconco™ 3970304

 View more versions of this product

Catalog No. 16-108-255

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Alert:

Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).


Requires : Work surface and base stand (sold separately).

Description

Description

  • Nominal downflow velocity of 45-65 fpm and aerodynamic air foil
  • Supply HEPA filter, 99.99% efficient on particles 0.3 Micron
  • ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
  • Variable speed built-in blower with motorized impeller and solid state control rated for 10A
  • Intrinsically-sound, negative pressure design
  • Minihelic pressure gauge and disposable prefilter
  • UV-resistant, glacier white and gray, powder-coated aluminum and steel construction
  • UV-opaque and resistant 6.3mm (0.25 in.) thick tempered safety glass sash and sides
  • Light and blower switches
  • Air diffuser and 23.8cm (9.4 in.) high sash opening
  • Ergonomic 10° angled and hinged sash that pivots upward, locks to a loading height of 50.8cm (20 in.) and has wiping seals to promote ISO Class 5 conditions
  • Two utility ports with iris openings and plugs
  • ETL Listing (115 volt, 60Hz models)

Order Info

Vertical Clean Benches can be ordered with bifold doors; specify when ordering. Guardian™ Airflow Monitor is offered as factory-installed option on all benches; specify when ordering.

Warranty

One year parts and service

Specifications

Specifications

Clean Bench
Fluorescent
36.8 in.
50/60 Hz
North America Plug
160 lb.
36 in.
Powder
5 A
93.47 cm
UV light, protection panel, timer and airflow monitor
230 V
73 kg
91.44 cm
SDS
Documents

Documents

Accessories
Promotions

Promotions