missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Purifier™ Vertical Clean Bench
Protect work from particulate contamination by choosing a Purifier™ Vertical Clean Bench.
$12928.93 - $20410.62
Specifications
Product Type | Clean Bench |
---|---|
Coating | Powder |
Lighting Type | Fluorescent |
Material | Powder-coated Aluminum and Steel |
Certifications/Compliance | CAN/CSA C22.2, UL, CE |
Alert:
Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).
Catalog Number | Mfr. No. | Plug Type | Amperage | Voltage | Includes | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Plug Type | Amperage | Voltage | Includes | Price | Quantity | ||
16-108-243
|
Labconco™
3970200 |
North America Plug | 5 A | 115 V | – |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-246
|
Labconco™
3970203 |
North America Plug | 5 A | 115 V | Airflow monitor |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-245
|
Labconco™
3970201 |
North America Plug | 5 A | 115 V | UV light, protection panel and timer |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-247
|
Labconco™
3970204 |
North America Plug | 5 A | 115 V | UV light, protection panel, timer and airflow monitor |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-252
|
Labconco™
3970300 |
North America Plug | 5 A | 230 V | – |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-256
|
Labconco™
3970320 |
No Plug | 3 A | 230 V | – |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-254
|
Labconco™
3970303 |
North America Plug | 5 A | 230 V | Airflow monitor |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-253
|
Labconco™
3970301 |
North America Plug | 5 A | 230 V | UV light, protection panel and timer |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-255
|
Labconco™
3970304 |
North America Plug | 5 A | 230 V | UV light, protection panel, timer and airflow monitor |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-260
|
Labconco™
3970400 |
North America Plug | 5 A | 115 V | – |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-261
|
Labconco™
3970401 |
North America Plug | 5 A | 115 V | UV light, protection panel and timer |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
16-108-263
|
Labconco™
3970404 |
North America Plug | 5 A | 115 V | UV light, protection panel, timer and airflow monitor |
N/A
|
N/A | ||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Nominal downflow velocity of 45-65 fpm and aerodynamic air foil
- Supply HEPA filter, 99.99% efficient on particles 0.3 Micron
- ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
- Variable speed built-in blower with motorized impeller and solid state control rated for 10A
- Intrinsically-sound, negative pressure design
- Minihelic pressure gauge and disposable prefilter
- UV-resistant, glacier white and gray, powder-coated aluminum and steel construction
- UV-opaque and resistant 6.3mm (0.25 in.) thick tempered safety glass sash and sides
- Light and blower switches
- Air diffuser and 23.8cm (9.4 in.) high sash opening
- Ergonomic 10° angled and hinged sash that pivots upward, locks to a loading height of 50.8cm (20 in.) and has wiping seals to promote ISO Class 5 conditions
- Two utility ports with iris openings and plugs
- ETL Listing (115 volt, 60Hz models)
Order Info
Vertical Clean Benches can be ordered with bifold doors; specify when ordering. Guardian™ Airflow Monitor is offered as factory-installed option on all benches; specify when ordering.
Warranty
One year parts and service
Specifications
Clean Bench | |
Fluorescent | |
CAN/CSA C22.2, UL, CE | |
28.6 in. | |
Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation | |
93.47 cm | |
Utility |
Powder | |
Powder-coated Aluminum and Steel | |
Glacier White, Gray | |
72.64 cm | |
36.8 in. | |
50/60 Hz |