missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Telescoping Base Stands with Casters

With 5 in. dia. (12.7cm) toe-locking casters

Supplier:  Labconco™ 3746712

 View more versions of this product

Catalog No. 16-101-124


Explore available promotions

Description

Description

  • Nonmarking polyurethane casters with bearings for quiet operation and added mobility
  • Length, 29 in. (74cm)

Order Info

  • Enclosures supported by mobile base stands will require special modifications to incorporate flexible ducting,
  • wiring and plumbing when applied.

Specifications

Specifications

3 ft. Enclosure
84 lb.
29 in.
5 in. dia. Lockable Nonmarking Polyurethane Casters
121.9 cm
ADA
34.5 to 27.5 in.
38 Kg
48 in.
73.66 cm
Stand
Videos
SDS
Documents

Documents

Promotions

Promotions