Labconco™ Test Tube Holders
Add-on accessory to the Lower Test Tube spindle rack
Supplier:  Labconco™ 4546300
 View more versions of this product
Catalog No. 10-359-155
Explore available promotions
Description
Description
  • Keeps tubes upright
  • Constructed of Type 304 stainless steel
Specifications
Specifications
Stainless Steel (304)
75
10.6 x 14.3 x 7.3 in. (26.9 x 36.2 x 18.4cm)
18.4 cm
26.9 cm
36.2 cm
Tubes (20 to 25 mm)
20 to 25mm Tubes
7.3 in.
10.6 in.
14.3 in.