Labconco™ Tube Holders for Fast-Freeze™ and Lyph-Lock™ Flasks
Holds tubes in upright position for freeze drying.
Supplier:  Labconco™ 7442700
 View more versions of this product
Catalog No. 10-030-159
Explore available promotions
Description
Description
Holds tubes in upright position for freeze drying.
Hold tubes in upright position inside 900mL or larger Fast-Freeze Flasks and 750mL or larger Lyph-Lock Flasks Stainless steel
Specifications
Specifications
Tube Holder
Stainless Steel
900ml or larger fast freeze™ flasks and 750ml or larger lyph lock flasks
8.33 cm
13.9 cm
0.5 kg
750, 900 mL
Hold tubes in upright position
3.5 in.
5 in.
0.5 lb.