Fisherbrand™ Laboratory Workstation with Black Phenolic Work Surface
Mobile Lab Workstations are furnished complete with shelving, electrical fixtures, and work surfaces. They include a heavy duty tubular steel base equipped with vibration isolation casters.
Supplier:  Fisherbrand™ FENOS3072P
 View more versions of this product
Catalog No. 06001085
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Adjustable steel shelving with wire lips
  • 125 V, 20 A electrical raceway with twist to lock plug
  • Table can have plumbing fixtures, data services, and suspended cabinetry added as needed
Specifications
Specifications
72 in.
213.4 cm
182.9 cm
SEFA 8
30 in.
76.2 cm
84 in.