missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lactic Acid (85% w/w/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific BPA1621

 View more versions of this product

Catalog No. A162-1



Explore available promotions

Description

Description

Specifications

Specifications

Lactic Acid
85% (w/w)
50-21-5,97-73-4
85.0 to 90.0%
CH3CH(OH)COOH
3265
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
2-hydroxypropanoic acid
612
85.0 to 90.0%
1.200g/cm³
0.020% max.
Viscous Liquid
2,8
18°C
122°C
C3H6O3
MFCD00004520
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
CC(O)C(O)=O
90.08
CHEBI:78320
Certified ACS
1 L
Glass Bottle

Chemical Identifiers

50-21-5
90.08
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
612
2-hydroxypropanoic acid
C3H6O3
MFCD00004520
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
CHEBI:78320
CC(O)C(O)=O
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions