Lactose, Monohydrate, Powder, ACS, Spectrum™ Chemical
Supplier:  Spectrum Chemical Mfg Cor L101525KG
 View more versions of this product
Catalog No. 18611184
Explore available promotions
Description
Description
Specifications
Specifications
64044-51-5
0.0003
C12H24O12
WSVLPVUVIUVCRA-RJMJUYIDSA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
ACS
1
Poly Bottle
2.5 kg
O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
360.31
5e-05
Chemical Identifiers
64044-51-5
360.31
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
C12H24O12
WSVLPVUVIUVCRA-RJMJUYIDSA-N
O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
Videos
SDS
Documents
Documents
Promotions
Promotions
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.