missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lactose, Monohydrate, Powder, ACS, Spectrum™ Chemical
$370.09 - $6394.38
Chemical Identifiers
CAS | 64044-51-5 |
---|---|
Molecular Formula | C12H24O12 |
Molecular Weight (g/mol) | 360.31 |
InChI Key | WSVLPVUVIUVCRA-RJMJUYIDSA-N |
IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate |
SMILES | O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18611183
|
Spectrum Chemical Mfg Cor
L1015500GM |
500 g | Poly Bottle |
Each for $370.09
|
|
||||
18611184
|
Spectrum Chemical Mfg Cor
L101525KG |
2.5 kg | Poly Bottle |
Each for $1,142.94
|
|
||||
18611185
|
Spectrum Chemical Mfg Cor
L101512KG |
12 kg | Poly Pail |
Each for $2,795.88
|
|
||||
18611186
|
Spectrum Chemical Mfg Cor
L101545KGBL |
45 kg | Fiber Drum |
Each for $6,394.38
|
|
||||
Chemical Identifiers
64044-51-5 | |
360.31 | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate |
C12H24O12 | |
WSVLPVUVIUVCRA-RJMJUYIDSA-N | |
O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O |
Specifications
64044-51-5 | |
0.0003 | |
C12H24O12 | |
WSVLPVUVIUVCRA-RJMJUYIDSA-N | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate | |
ACS |
1 | |
Poly Bottle | |
500 g | |
O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O | |
360.31 | |
5e-05 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.