missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lamivudine 98.0+%, TCI America™

Supplier:  TCI America L02171G

 View more versions of this product

Catalog No. L02171G


Explore available promotions

Description

Description

Specifications

Specifications

Lamivudine
177°C
C8H11N3O3S
1 g
JTEGQNOMFQHVDC-NKWVEPMBSA-N
4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
60825
229.25
Crystalline Powder
134678-17-4
White-Yellow
MFCD00869739
lamivudine, epivir, zeffix, heptovir, epivir-hbv, hepitec, heptodin, 2',3'-dideoxy-3'-thiacytidine, lamivir, 3tc
C1C(OC(S1)CO)N2C=CC(=NC2=O)N
229.254
CHEBI:63577
≥98.0% (HPLC,T)

Chemical Identifiers

134678-17-4
229.254
JTEGQNOMFQHVDC-NKWVEPMBSA-N
60825
4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
C8H11N3O3S
MFCD00869739
lamivudine, epivir, zeffix, heptovir, epivir-hbv, hepitec, heptodin, 2',3'-dideoxy-3'-thiacytidine, lamivir, 3tc
CHEBI:63577
C1C(OC(S1)CO)N2C=CC(=NC2=O)N
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UW7361333

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions