missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lamivudine 98.0+%, TCI America™
$29.41 - $147.06
Chemical Identifiers
CAS | 134678-17-4 |
---|---|
Molecular Formula | C8H11N3O3S |
Molecular Weight (g/mol) | 229.254 |
MDL Number | MFCD00869739 |
InChI Key | JTEGQNOMFQHVDC-NKWVEPMBSA-N |
Synonym | lamivudine, epivir, zeffix, heptovir, epivir-hbv, hepitec, heptodin, 2',3'-dideoxy-3'-thiacytidine, lamivir, 3tc |
PubChem CID | 60825 |
ChEBI | CHEBI:63577 |
IUPAC Name | 4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one |
SMILES | C1C(OC(S1)CO)N2C=CC(=NC2=O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
L0217100MG
|
TCI America
L0217100MG |
100 mg |
Each for $29.41
|
|
|||||
L02171G
|
TCI America
L02171G |
1 g |
Each for $147.06
|
|
|||||
Chemical Identifiers
134678-17-4 | |
229.254 | |
JTEGQNOMFQHVDC-NKWVEPMBSA-N | |
60825 | |
4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one |
C8H11N3O3S | |
MFCD00869739 | |
lamivudine, epivir, zeffix, heptovir, epivir-hbv, hepitec, heptodin, 2',3'-dideoxy-3'-thiacytidine, lamivir, 3tc | |
CHEBI:63577 | |
C1C(OC(S1)CO)N2C=CC(=NC2=O)N |
Specifications
134678-17-4 | |
White-Yellow | |
MFCD00869739 | |
lamivudine, epivir, zeffix, heptovir, epivir-hbv, hepitec, heptodin, 2',3'-dideoxy-3'-thiacytidine, lamivir, 3tc | |
C1C(OC(S1)CO)N2C=CC(=NC2=O)N | |
229.254 | |
CHEBI:63577 | |
≥98.0% (HPLC,T) | |
Lamivudine |
177°C | |
C8H11N3O3S | |
100 mg | |
JTEGQNOMFQHVDC-NKWVEPMBSA-N | |
4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one | |
60825 | |
229.25 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : UW7361333
TSCA : No
Recommended Storage : Freezer