Learn More
Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO), Thermo Scientific Chemicals
Form: Approx.0.20g/mm, Cast rod, diameter is approximate
Supplier: Thermo Scientific Chemicals 040292DJ
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver-White | |
Rare Earth Oxides Content | |
Silvery-white rod | |
99.9% (REO) | |
MFCD00011066 | |
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
[La] | |
138.91 | |
CHEBI:33336 | |
Cast Rod |
Lanthanum | |
7439-91-0 | |
6.17g/cm3 at 20°C | |
La | |
50 mm | |
FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
lanthanum | |
23926 | |
Mylar under Argon |
Chemical Identifiers
7439-91-0 | |
138.91 | |
FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
23926 | |
lanthanum |
La | |
MFCD00011066 | |
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
CHEBI:33336 | |
[La] |
Safety and Handling
Keep container tightly closed and dry. This material and/or its container must be disposed of as hazardous waste.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : DOT Class: 4.3, Packaging Group: II
EINECSNumber : 231-099-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only