missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO), Thermo Scientific Chemicals
Form: Approx.0.20g/mm, Cast rod, diameter is approximate
$297.67 - $1501.12
Chemical Identifiers
CAS | 7439-91-0 |
---|---|
Molecular Formula | La |
Molecular Weight (g/mol) | 138.91 |
MDL Number | MFCD00011066 |
InChI Key | FZLIPJUXYLNCLC-UHFFFAOYSA-N |
Synonym | in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation |
PubChem CID | 23926 |
ChEBI | CHEBI:33336 |
IUPAC Name | lanthanum |
SMILES | [La] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA40292HF
|
Thermo Scientific Chemicals
040292HF |
10 mm |
Each for $297.67
|
|
|||||
AA40292DH
|
Thermo Scientific Chemicals
040292DH |
25 mm |
Each for $427.33
|
|
|||||
AA40292DJ
|
Thermo Scientific Chemicals
040292DJ |
50 mm |
Each for $767.00
|
|
|||||
AA40292DM
|
Thermo Scientific Chemicals
040292DM |
100 mm |
Each for $1,501.12
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-91-0 | |
138.91 | |
FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
23926 | |
lanthanum |
La | |
MFCD00011066 | |
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
CHEBI:33336 | |
[La] |
Specifications
Silver-White | |
Silvery-white rod | |
La | |
10 mm | |
FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
lanthanum | |
23926 | |
Cast Rod |
7439-91-0 | |
99.9% (REO) | |
MFCD00011066 | |
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
[La] | |
138.91 | |
CHEBI:33336 | |
Lanthanum, Rare Earth Oxides Content |
Safety and Handling
Keep container tightly closed and dry. This material and/or its container must be disposed of as hazardous waste.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : DOT Class: 4.3, Packaging Group: II
EINECSNumber : 231-099-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only