Brady™ LASERTAB™ Laser Printable Labels, B-747 Polyester
Designed for use with laser printers. Brady LASERTAB Laser Printable Labels are made of B-747 polyester.
$242.62 - $746.19
Specifications
Adhesive Type Acrylic, Permanent
Finish Matte
Material Polyester (B-747)
Product Type Laser Printable Label
Thickness (English) 0.0037 in.
View More Specs
Products 21
Catalog Number Mfr. No. Size (Metric) Size (English) Color Price Quantity  
Catalog Number Mfr. No. Size (Metric) Size (English) Color Price Quantity  
19-112-756
View Documents Promotion Details
Brady™
LAT117475
47.6 x 9.52 mm (W x H) 1.875 x 0.375 in. (W x H) White
Pack of 5000 Each for $484.26
19-112-757
View Documents Promotion Details
Brady™
LAT1274725
76.2 x 9.52 mm (W x H) 3 x 0.375 in. (W x H) Yellow
Pack of 2500 Each for $361.42
19-112-758
View Documents Promotion Details
Brady™
LAT1374725
47.6 x 21.1 mm (W x H) 1.875 x 0.833 in. (W x H) White
Pack of 2500 Each for $426.38
19-112-759
View Documents Promotion Details
Brady™
LAT1374725YL
47.6 x 21.1 mm (W x H) 1.875 x 0.833 in. (W x H) Yellow
Pack of 2500 Each for $559.39
19-112-760
View Documents Promotion Details
Brady™
LAT1474715
47.6 x 33.7 mm (W x H) 1.875 x 1.33 in. (W x H) White
Pack of 1500 Each for $461.91
19-112-761
View Documents Promotion Details
Brady™
LAT174710
7.62 x 6.35 mm (W x H) 0.3 x 0.25 in. (W x H) White
Pack of 10000 Each for $342.11
19-112-775
View Documents Promotion Details
Brady™
LAT217471YL
63.5 x 25.4 mm (W x H) 2.5 x 1 in. (W x H) Yellow
Pack of 1000 Each for $456.85
19-112-774
View Documents Promotion Details
Brady™
LAT217471
63.5 x 25.4 mm (W x H) 2.5 x 1 in. (W x H) White
Pack of 1000 Each for $242.62
19-112-780
View Documents Promotion Details
Brady™
LAT2574710
12.7 x 11.09 mm (W x H) 0.5 x 0.437 in. (W x H) Yellow
Pack of 10000 Each for $323.84
19-112-781
View Documents Promotion Details
Brady™
LAT2674710
6.35 x 6.35 mm (W x H) 0.25 x 0.25 in. (W x H) White
Pack of 10000 Each for $342.11
19-112-782
View Documents Promotion Details
Brady™
LAT274710
9.52 x 9.52 mm (W x H) 0.375 x 0.375 in. (W x H) White
Pack of 10000 Each for $316.75
19-112-783
View Documents Promotion Details
Brady™
LAT2874725SH
215.9 x 279.4 mm (W x H) 8.5 x 11 in. (W x H) White
Pack of 25 Each for $258.12
19-112-784
View Documents Promotion Details
Brady™
LAT28747Y25SH
215.9 x 279.4 mm (W x H) 8.5 x 11 in. (W x H) Yellow
Pack of 25 Each for $287.29
19-112-786
View Documents Promotion Details
Brady™
LAT2974710
16.5 x 5.08 mm (W x H) 0.65 x 0.2 in. (W x H) White
Pack of 10000 Each for $353.29
19-112-787
View Documents Promotion Details
Brady™
LAT2974710YL
16.5 x 5.08 mm (W x H) 0.65 x 0.2 in. (W x H) Yellow
Pack of 10000 Each for $381.71
19-112-795
View Documents Promotion Details
Brady™
LAT374710
16.5 x 6.35 mm (W x H) 0.65 x 0.25 in. (W x H) White
Pack of 10000 Each for $353.29
19-112-800
View Documents Promotion Details
Brady™
LAT474710
20.3 x 6.35 mm (W x H) 0.8 x 0.25 in. (W x H) White
Pack of 10000 Each for $411.15
19-112-802
View Documents Promotion Details
Brady™
LAT574710
20.3 x 12.7 mm (W x H) 0.8 x 0.5 in. (W x H) White
Pack of 10000 Each for $684.27
19-112-806
View Documents Promotion Details
Brady™
LAT774710
25.4 x 12.7 mm (W x H) 1 x 0.5 in. (W x H) Yellow
Pack of 10000 Each for $687.30
19-112-807
View Documents Promotion Details
Brady™
LAT774710YL
25.4 x 12.7 mm (W x H) 1 x 0.5 in. (W x H) White
Pack of 10000 Each for $746.19
19-112-811
View Documents Promotion Details
Brady™
LAT974710
38.1 x 6.35 mm (W x H) 1.5 x 0.25 in. (W x H) White
Pack of 10000 Each for $476.14
Description
Description
  • Blank, printable polyester labels for Laser Printers
  • CSA Accepted material when printed with designated laser printer toners
Specifications
Specifications
Acrylic, Permanent
Polyester (B-747)
0.0037 in.
Matte
Laser Printable Label
SDS
missing translation for 'documents'
missing translation for 'documents'