missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) bromide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1940636

 View more versions of this product

Catalog No. AAA1940636


Explore available promotions

Description

Description

It finds its uses as a catalyst to make polyesters, a filler for polypropylene (flame-resistant), and in glass optical waveguides. It also finds ints use in laboratories and metallurgy. it is used as acryl amide monomer light aggregation function of catalysts and can be used for photoelectric material and heat power semiconductor materials; it is also used as analytical reagent, additive and in the preparation of massicot pigment; it is also used as pigment ingredient and used for printing and dyeing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as a catalyst to make polyesters, a filler for polypropylene (flame-resistant), and in glass optical waveguides. It also finds ints use in laboratories and metallurgy. it is used as acryl amide monomer light aggregation function of catalysts and can be used for photoelectric material and heat power semiconductor materials; it is also used as analytical reagent, additive and in the preparation of massicot pigment; it is also used as pigment ingredient and used for printing and dyeing industry.

Solubility
Soluble in water at 20°C 5g/L. Insoluble in ethanol. Solubility in N,N-DMF is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

916°C
373°C
500 g
PbBr2
UN2291
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
λ²-lead(2+) dibromide
24831
≥98%
6.66
Lead(II) bromide
10031-22-8
Br2Pb
MFCD00011156
14,5403
Soluble in water at 20°C 5g/L. Insoluble in ethanol. Solubility in N,N-DMF is almost transparent.
[Br-].[Br-].[Pb++]
367.00
367.01
Odorless

Chemical Identifiers

10031-22-8
367.00
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
24831
Br2Pb
MFCD00011156
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
[Br-].[Br-].[Pb++]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c

H302+H332-H360Df-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

EINECSNumber : 233-084-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only