missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) bromide, 98+%, Thermo Scientific Chemicals

$85.14 - $226.73

Chemical Identifiers

CAS 10031-22-8
Molecular Formula Br2Pb
Molecular Weight (g/mol) 367.00
MDL Number MFCD00011156
InChI Key ZASWJUOMEGBQCQ-UHFFFAOYSA-L
Synonym lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
PubChem CID 24831
SMILES [Br-].[Br-].[Pb++]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1940622
View Documents
Thermo Scientific Chemicals
A1940622
100 g
Each for $85.14
 
AAA1940636
View Documents
Thermo Scientific Chemicals
A1940636
500 g
Each for $226.73
 
Description

Description

It finds its uses as a catalyst to make polyesters, a filler for polypropylene (flame-resistant), and in glass optical waveguides. It also finds ints use in laboratories and metallurgy. it is used as acryl amide monomer light aggregation function of catalysts and can be used for photoelectric material and heat power semiconductor materials; it is also used as analytical reagent, additive and in the preparation of massicot pigment; it is also used as pigment ingredient and used for printing and dyeing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as a catalyst to make polyesters, a filler for polypropylene (flame-resistant), and in glass optical waveguides. It also finds ints use in laboratories and metallurgy. it is used as acryl amide monomer light aggregation function of catalysts and can be used for photoelectric material and heat power semiconductor materials; it is also used as analytical reagent, additive and in the preparation of massicot pigment; it is also used as pigment ingredient and used for printing and dyeing industry.

Solubility
Soluble in water at 20°C 5g/L. Insoluble in ethanol. Solubility in N,N-DMF is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

10031-22-8
367.00
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
24831
Br2Pb
MFCD00011156
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
[Br-].[Br-].[Pb++]

Specifications

916°C
10031-22-8
Br2Pb
MFCD00011156
14,5403
Soluble in water at 20°C 5g/L. Insoluble in ethanol. Solubility in N,N-DMF is almost transparent.
[Br-].[Br-].[Pb++]
367.00
367.01
Odorless
Lead(II) bromide
373°C
100 g
PbBr2
UN2291
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
dibromolead
24831
≥98%
6.66
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P312-P314-P330-P501c

H302+H332-H360Df-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

EINECSNumber : 233-084-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only